Learn More
Medchemexpress LLC PKI-beta Human His 50ug

Supplier: Medchemexpress LLC HYP7120950UG
PKI-beta is a recombinant human protein that functions as a potent, competitive inhibitor of cAMP-dependent protein kinase (PKA). Expressed in Escherichia coli with an N-terminal 6xHis tag, it is supplied as a filtered, glycerol-containing buffered solution and is purified for use in in vitro biochemical assays that require selective inhibition of PKA catalytic activity.
- Recombinant human isoform-1 (M1-K78) for biological relevance.
- Potent competitive inhibition of PKA catalytic subunit.
- N-terminal 6xHis tag for affinity purification or detection.
- >95% purity confirmed by reducing SDS-PAGE.
- Stabilized glycerol-containing formulation for improved shelf life.
- Available in small-scale sizes (10 μg, 50 μg, 100 μg) for assay flexibility.
- Recommended storage at -80°C; avoid repeated freeze-thaw cycles.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.