Learn More
Medchemexpress LLC PKLR Human HEK293 50ug
SDP
Supplier:  Medchemexpress LLC HYP7121050UG
Encompass_Preferred
Recombinant human pyruvate kinase (PKLR) produced in HEK293 cells and supplied as a filtered, stabilized solution with a C-terminal 6xHis tag for research use. The protein is >95% pure by reducing SDS-PAGE, has an approximate molecular weight of 62.0 kDa, and is provided in microgram-scale quantities for biochemical and assay applications.
  • Expressed in HEK293 cells with a C-terminal 6xHis tag.
  • Purity greater than 95% as determined by reducing SDS-PAGE.
  • Approximate molecular weight 62.0 kDa.
  • Supplied in a stabilized formulation with glycerol, trehalose, and buffering salts.
  • Stored at -80°C and shipped on dry ice; avoid repeated freeze-thaw.
  • Available in small microgram quantities suitable for biochemical assays and reagent use.
Catalog No. 50-001-96957