Learn More
Selleck Chemical LLC PKM2 inhibitor(compound 3k) 5mg 94164-88-2
SDP
Supplier:  Selleck Chemical LLC S86165MG
Encompass_Preferred
PKM2 inhibitor(Compound 3K) displays PKM2 inhibitory activity with the IC50 value of 2.95 ?M. The IC50 value for PKM1 is 4-5-fold higher than that for PKM2. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8073