Learn More
Medchemexpress LLC PKR-IN-C16 | 608512-97-6 | 99.7% | 268.29 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY13977A1MG
Encompass_Preferred
PKR-IN-C16 (Compound C16) is a specific double-stranded RNA-dependent protein kinase (PKR) inhibitor. It exhibits promising neuroprotective properties, can rescue acute brain lesions, and helps unlock translation blockade induced by PKR in primary neuronal cultures.
  • Specific double-stranded RNA-dependent protein kinase (PKR) inhibitor
  • Exhibits neuroprotective properties
  • Rescues acute brain lesions
  • Protects against neuronal cell death induced by endoplasmic reticulum stress
  • Prevents PKR-phosphorylation and activation of caspase-3
  • Reduces phosphorylated PKR levels
  • Prevents PKR-induced neuronal loss and inflammatory response
Catalog No. 50-003-51261