Learn More
Medchemexpress LLC PKR-IN-C16 (Compound C16) | 608512-97-6 | 99.7% | 268.29 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY13977A10MM/1ML
Encompass_Preferred
PKR-IN-C16 (Compound C16) is a specific inhibitor of double-stranded RNA-dependent protein kinase (PKR). It demonstrates promising neuroprotective properties and has the ability to rescue acute brain lesions.
  • Specific double-stranded RNA-dependent protein kinase (PKR) inhibitor.
  • Exhibits promising neuroprotective properties.
  • Can rescue acute brain lesions.
Catalog No. 50-003-49877