Learn More
Medchemexpress LLC Pkr-in-c51 | 1314594-23-4 | MFCD34676504 | 99.8% | 367.45 | C23H21N5 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY118131100MG
Encompass_Preferred
Selective small-molecule inhibitor of protein kinase R (PKR) intended for laboratory research. Supplied as a light yellow solid and typically provided in a 100 MG package. Reported purity 99.79%, molecular weight 367.45 g/mol, and formula C23H21N5. Documentation includes a datasheet, certificate of analysis, and safety data sheet. Store at 4°C protected from light; in solution store at -80°C for long-term stability.
  • Selective inhibitor of protein kinase R (PKR) for biochemical and cellular studies.
  • Reported purity 99.79% suitable for research applications.
  • Light yellow solid; available in 100 MG packaging and DMSO solution formats.
  • Includes datasheet, COA, and SDS for quality and safety information.
  • Storage instructions supplied; store at 4°C protected from light, in solution store at -80°C.
Catalog No. 50-002-26524