Learn More
Medchemexpress LLC Pkr-in-c51 (compound 51) | 1314594-23-4 | 99.8% | 367.45 g/mol | C23H21N5 | 25 MG

Supplier: Medchemexpress LLC HY11813125MG
PKR-IN-C51 (compound 51) is an ATP-competitive inhibitor of double-stranded RNA-activated protein kinase (PKR) that suppresses intracellular PKR activation in primary mouse macrophages. It has an IC50 of 9 μM and is supplied as a solid or as a DMSO solution for research use only.
- ATP-competitive PKR inhibitor; IC50 9 μM.
- Molecular weight 367.45 g/mol; formula C23H21N5.
- High purity (≈99.8%).
- Available as solid (25 MG) and as 10 mM solution in DMSO.
- Storage: solid at 4°C protected from light; in solvent store at -80°C (6 months) or -20°C (1 month).
- For research use only; not for human or clinical use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.