Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Pksi-527 | 128837-71-8 | MFCD01745772 | 98.1% | 473.99 | C25H32ClN3O4 | 25MG
SDP

Supplier:  Medchemexpress LLC HY12313325MG

Encompass_Preferred

PKSI-527 is a research compound that functions as a highly selective plasma kallikrein inhibitor. It has been reported to suppress collagen-induced arthritis by modulating the kallikrein-kinin system and is provided with sufficient analytical purity for biological research applications.

  • Selective plasma kallikrein inhibitor.
  • Reported to suppress collagen-induced arthritis in preclinical models.
  • Purity 98.08% as supplied for research use.
  • Chemical formula C25H32ClN3O4, molecular weight 473.99.
  • Soluble in DMSO at 100 mg/mL (≈210.97 mM); ultrasonic agitation recommended.
  • Intended for use in biochemical and pharmacological studies of the kallikrein-kinin pathway.

Catalog No. 50-002-38393


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.