Learn More
Medchemexpress LLC PKSI-527 | 128837-71-8 | MFCD01745772 | 98.1% | 473.99 g/mol | C25H32ClN3O4 | 5MG
SDP
Supplier:  Medchemexpress LLC HY1231335MG
Encompass_Preferred
PKSI-527 is a small-molecule, highly selective plasma kallikrein inhibitor used in preclinical research. It has been shown to suppress collagen-induced arthritis by modulating the kallikrein-kinin system. The compound is supplied as a powder and is also available as a 10 mM solution in DMSO to support various in vitro and in vivo study formats.
  • Highly selective plasma kallikrein inhibition for target validation and mechanistic studies.
  • Demonstrated activity in collagen-induced arthritis models.
  • Available as a powder and as a 10 mM solution in DMSO for experimental flexibility.
  • Multiple package sizes support small-scale screening and larger preclinical studies.
  • Characterized by molecular formula C25H32ClN3O4 and molecular weight 473.99 g/mol.
  • Typical assay purity reported around 98% by HPLC; certificate of analysis available per batch.
Catalog No. 50-002-38704