Learn More
Enzo Life Sciences PKSI-527 (25mg). CAS: 128837-71-8
SDP
Supplier:  Enzo Life Sciences BMLPI1470025
Encompass_Preferred
Highly selective plasma kallikrein inhibitor (Ki = 0.81 µM). Can be used for affinity purification of kalikrein. Suppresses collagen-induced arthritis in a mouse model. Synergizes with NO donors producing antithrombotic effects ex vivo. Alternative name: trans-4-Aminomethylcyclohexanecarbonyl-phenylalanyl-4-carboxymethylanilide. Purity: ≥95% (HPLC). Solubility: Soluble in water (5 mg/ml). Long Term Storage: -20°C.
Catalog No. 50-201-1490