Learn More
BEZWADA BIOMEDICAL LLC Poly(L-lactide-co-caprolactone), L-lactide:caprolactone 60:40,viscosity 1.0 to 2.0 dl/g, 5.0 GM
SDP
Supplier:  BEZWADA BIOMEDICAL LLC 3164
Encompass_Preferred
The presence of caprolactone repeat units in the copolymer reduces the degree of close packing between lactide repeat units leaving the polymer with nice elastic properties while maintaining a relatively slow degradation profile. This allows PLA-PCL copolymers to be used in drug delivery and medical device coating applications for controlled release applications. Given below is the collection of our Caprolactone/L-Lactide copolymers. The polymers are used in medical device, drug delivery and 3D printing.
Catalog No. 50-190-4467