Learn More
BEZWADA BIOMEDICAL LLC Poly(L-lactide-co-glycolide), L-lactide:glycolide 65:35,viscosity 0.50 to 1.0 dl/g, 5.0 GM
SDP
Supplier:  BEZWADA BIOMEDICAL LLC 3263
Encompass_Preferred
The complete absorption of L(-)-PLA requires more than 2 years and is much slower than that of DL-PLA. In order to decrease the absorption times of implantable devices incorporating L(-)-lactide, copolymers of L(-)-lactide and dl-lactide with glycolide have been prepared. The polymers are used in medical devices and drug delivery.
Catalog No. 50-190-4466