Learn More
Aviva Systems Biology Corp PLAGL1 Antibody - N-terminal region (ARP38852_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38852_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against PLAGL1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human PLAGL1. Alternate names: ZAC, LOT1, ZAC1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-56821