Learn More
Electron Microscopy Sciences Tripod Polisher Plain "L" Bracket
SDP
Supplier:  Electron Microscopy Sciences 5011620
Encompass
The Plain L-Brackets (#50116-20): Can be used in place of the other ‘L' bracket to polish the edge of the samples to the area of interest prior to polishing the wedge. - For use with Tripod Polisher Model 590.
Catalog No. 50-980-867