Learn More
INTACT GENOMICS INC FastAmp Plant Cell Lysis Solution 20 mL
Supplier:  INTACT GENOMICS INC 4611
Encompass
Intact Genomics proprietary FastAmp DNA, RNA and Cell Solution (former name: FastAmp Viral and Cell Solution for COVID-19 Testing) can be utilized for saliva/swab samples resulting in viral/cell lysis, and significantly simplifying this process. This solution streamlines specimen collection and DNA or RNA preparation processes including sample collection, transport, maintenance, viral/cell lysis, and viral RNA stabilization. After using the solution, the lysed DNA or RNA may be directly used in PCR, RT-qPCR, RT-isothermal DNA amplification, without the need of an RNA extraction step.Intact Genomics FastAmp DNA, RNA and Cell Solution can be used various sample and cell types stabilizing and prepping DNA or RNA for subsequent PCR or cDNA synthesis, saving your team significant time and resources.
Catalog No. 50-000-82935