Learn More
LUNA NANOTECH INC 96-Well Plant Genomic Dna Extraction Plus Kit, 384 Preps
Supplier:  LUNA NANOTECH INC NKP262384
Encompass_Preferred
PuroSPIN-96™ Plant Genomic DNA Purification Plus Kit, 96-Well Plate Format, 384 Preps
The PuroSPIN-96™ Plant Genomic DNA Purification Plus Kit is a high throughput method designed to purify genomic DNA from variety of plant species and tissues types. The kit greatly simplifies the purification process by incorporating a convenient and easy-to-use extraction technology based on nucleic acid binding to a silica membrane assembled in each well of a 96-well plate. Use of the silica membrane for purififcation, eliminates the toxic phenol-chloroform extraction or time consuming alcohol precipitation.
In comparison to our standard PuroSPIN-96™ Plant Genomic DNA Purificatio Kit, the Plus kit adds an additional 96-well plate filtration step that removes the insoluble and non-digested fraction from the lysate before loading it onto the 96-well silica spin column plate. The 96-well plate filtration replaces precipitation of insoluble fraction by centrifuga
Catalog No. 50-256-6916