Learn More
Percival Scientific Plant Growth Chamber, E36VLH11, 29.7 cu. ft., vertical SciWhite with red LEDs, 5 tiers, 7-44° C ±0.5°, IntellusUltra controller, touchscreen, advanced ultrasonic humidifier
SDP
Supplier:  Percival Scientific E36VLH11
Encompass_Preferred
The E36VLH11 growth chamber is frequently used for plant production and growing Arabidopsis. Vertical SciWhite with red LEDs are dimmable from 10-100% output with up to 515 µmoles/m2/s irradiance @ 6". IntellusUltra controller, touchscreen, advanced additive ultrasonic humidifier. Interior/exterior of smooth white galvanized steel w/ foamed-in-place non-CFC insulation and stainless-steel floor for corrosion resistance. Air circulation inside chamber is from a specifically designed adjustable air diffuser. Five epoxy coated steel wire shelves adjustable in ½” increments. Self-contained air-cooled condensing unit w/ hot gas bypass system for continuous compressor operation extends life, ensuring precise temperature control by cycling refrigerant and hot gas to coil; also prolongs life of compressor, and eliminates risk of ice build up in coil. Top mounted refrigeration system allows easy access for maintenance and cleaning. Contact sales@percival-scientific.com for more options.
Catalog No. NC3795089
Only null left
Add to Cart
Request a Quote