Learn More
Percival Scientific Plant Growth Chamber, E75L1H12, 76.1 cu. ft., SciWhite with red LEDs, 1 tier, 7-44° C ±0.5°, IntellusUltra controller, touchscreen, advanced ultrasonic humidifier and dehumidifier
SDP
Supplier:  Percival Scientific E75L1H12
Encompass_Preferred
The E75L1H12 growth chamber is frequently used for cereals, citrus, grasses and other plants that require high light intensity and higher growth height. SciWhite with red LEDs are dimmable from 10-100% output with up to 1360 µmoles/m2/s irradiance @ 6". IntellusUltra controller, touchscreen, advanced ultrasonic humidifier and dehumidifier. Interior / exterior of smooth white galvanized steel with foamed-in-place non-CFC insulation and a stainless-steel floor for corrosion resistance. Air circulation inside chamber is from a specifically designed adjustable air diffuser. Two white epoxy coated steel wire shelves adjustable in ½” increments. Self-contained air-cooled condensing unit with hot gas bypass system for continuous compressor operation extends life, ensuring precise temperature control by alternately cycling refrigerant and hot gas to coil; this also prolongs life of compressor, and eliminates risk of ice build up in coil. Contact sales@percival-scientific.com for more options.
Catalog No. NC3795589
Add to Cart
Request a Quote