Learn More
Medchemexpress LLC Plasma kallikrein-IN-4 | 1357954-34-7 | C22H23N7O | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY153115
Encompass_Preferred
Plasma kallikrein-IN-4 (Example 153) is a plasma kallikrein inhibitor with an IC50 of 0.016 μM against human plasma kallikrein.
  • High purity
  • Potent inhibitor
  • Documented support: Data Sheet, COA, SDS, and Handling Instructions are available
  • Detailed solubility information for in vitro use
  • Recommended storage conditions
  • Intended for research use only
Catalog No. 50-003-74719