Learn More
Invent Biotechnologies Inc Minute™ Plasma Membrane-Derived Lipid Raft Isolation Kit - Spin Column-Based (20 preps)
Supplier:  Invent Biotechnologies Inc LR042
Encompass
Lipid rafts are small membrane domains containing high level of cholesterol and sphingolipids. Lipid rafts have been found in plasma membrane (PM) and internal organellar membranes such as mitochondria-associated membrane (MAMs) and endoplasmic reticulum. Lipid rafts are implicated in numerous cellular processes such as signal transduction, membrane trafficking and protein sorting. Lipid-modified proteins and some transmembrane proteins are concentrated in the rafts while other proteins are excluded. Lipid rafts are also found to be associated with Na+/K+ ATPase on PM. More information: https://inventbiotech.com/products/minute%E2%84%A2-plasma-membrane-derived-lipid-raft-isolation-kit-20-preps?_pos=1&_sid=89f0094ad&_ss=r*For total lipid raft isolation, please refer to MinuteTM Total Lipid Raft Isolation Kit under Cat # LR-039 (Invent Biotechnologies, Inc.).
Catalog No. NC1705759