Learn More
LUNA NANOTECH INC Plasmid Miniprep Kit, 96-Well, 384 Preps
Supplier:  LUNA NANOTECH INC NKP111384
Encompass_Preferred
PuroSPIN-96™ Plasmid Miniprep Kit, 96-Well Plate Format, 384 Preps
The PuroSPIN™ 96 Plasmid Miniprep Kit is designed to provide a simple and easy-to-use method for large-scale extraction and purification of plasmid DNA.
This purification technology is based on plasmid DNA binding to 96-well silica membrane purification plates, which allows for efficient removal of genomic DNA, protein, salt, and other contaminants. Silica purification plates provide a faster, simpler, and safer alternative to traditional nucleic acid purification techniques such as phenol-chloroform extraction.
The PuroSPIN™ buffer formulations produce highly purified plasmid DNA for downstream applications, such as transfection, enzymatic digestion, or PCR.
Features:
- Easily purify plasmids from bacterial cultures
- High throughput sample processing using 96-well silica membrane purification plates
- Optimized buffer formulations t
Catalog No. 50-256-6919