Learn More
LUNA NANOTECH INC Plasmid Miniprep Plus Kit, 96-Well, 384 Preps
Supplier:  LUNA NANOTECH INC NKP211384
Encompass_Preferred
PuroSPIN-96™ Plasmid Miniprep Plus Kit, 96-Well Plate Format, 384 Preps
The PuroSPIN-96™ Plasmid Miniprep Kit is designed to provide a simple and easy-to-use method for large-scale extraction and purification of plasmid DNA.
This purification technology is based on plasmid DNA binding to 96-well silica membrane purification plates, which allows for efficient removal of genomic DNA, protein, salt, and other contaminants. Silica purification plates provide a faster, simpler, and safer alternative to traditional nucleic acid purification techniques such as phenol-chloroform extraction.
In comparison to our standard PuroSPIN-96™ Plasmid Miniprep Kit, the Plus kit adds an additional filtration step that removes the precipitated SDS-protein complexes, genomic DNA, carbohydrates and other contaminants from the lysate before loading it onto the silica spin plates. This prevents contaminating materials from binding to and blocking parts of t
Catalog No. 50-256-6918