Learn More
LUNA NANOTECH INC Plasmid Miniprep Plus Kit, 200 Preps
Supplier:  LUNA NANOTECH INC NK211200
Encompass_Preferred
PuroSPIN™ Plasmid Miniprep Plus Kit, 200 Preps
The PuroSPIN™ Plasmid Miniprep Plus Kit is designed to provide a simple and easy-to-use method for extraction and purification of plasmid DNA from cultured bacterial cells. The PuroSPIN™ buffer formulations produce highly purified plasmid DNA for downstream applications.
In comparison to our standard PuroSPIN™ Plasmid Miniprep Kit, the Plus kit adds an additional filtration step that removes the precipitated SDS-protein complexes, genomic DNA, carbohydrates and other contaminants from the lysate before loading it onto the silica spin column. This prevents contaminating materials from binding to and blocking parts of the silica membrane, leading to an increased plasmid DNA yield and higher purity of the final product. This is particularly benficial when workign with large samples that contain a lot of starting material that needs to be processed.
Features:
- Easily purify pl
Catalog No. 50-256-6940