Learn More
Cell Biolabs Inc pMX Retroviral Cloning Vector, 10 µg
Supplier:  Cell Biolabs Inc RTV010
Encompass_Preferred
Clone your gene of interest into Cell Biolabs' pMX Retroviral Vector and then package into a retroviral particle using a retroviral packaging cell or co-transfect with retroviral packaging constructs into 293T cells.
Catalog No. NC0018708