Learn More
Electron Microscopy Sciences Plastic Canoe Small
SDP
Supplier:  Electron Microscopy Sciences 7004310
Encompass_Preferred
Flexible dishes for handling liquids or solids. Ideal for controlled pour-off. These Anti-Static Canoes with contoured sides provide a convenient non-slip gripping surface. As well, they allow for more accurate weighing and reading of a sample. The canoe shape enables pouring of weighed samples.
Size: 1 3/8"
Catalog No. 50-189-9213