Learn More
Electron Microscopy Sciences Plastic Forceps with Jaw Grips 6/pk
SDP
Supplier:  Electron Microscopy Sciences 72971
Encompass_Preferred
All plastic. These forceps are excellent for handling hot, cold, corrosive or contaminated objects. Made from blue poly-propylene with interlocking teeth. A catch locks tips tightly for use as a clamp. Scissor type handles with double action hinges makes this forcep a useful cryo tool, and great for handling stubs.
Catalog No. 50-278-79