Learn More
MSC USA Sealing ZUSA-HT-666 White Teflon PTFE Tube; 1/2" Inside Diameter, 5/8" Outside Diameter, 1/16" Wall Thickness; Maximum Working Pressure (psi): 100
Supplier:  MSC 29972502
Encompass
USA Sealing ZUSA-HT-666 White Teflon PTFE Tube; 1/2" Inside Diameter, 5/8" Outside Diameter, 1/16" Wall Thickness; Maximum Working Pressure (psi): 100
Catalog No. NC2027705