Learn More
Grainger Plastic Strapping Kit 1/2 PK25
Supplier:  Grainger 16P025
Encompass_Preferred
The Grainger 16P025 is a 1/4 HP, 115V, 60Hz, 1-phase, open drip-proof electric motor with a 48 frame size. It has a 1075 RPM rated speed and features a sleeve bearing with permanently lubricated construction. This motor has a continuous duty rating and includes a 6 foot power cord with 3 prong plug. The enclosure is thermally protected for overload conditions. It can be used for driving fans, blowers, pumps and other light industrial applications requiring up to 1/4 HP.
Catalog No. 50-276-4590