Learn More
Thomas Scientific 700mm Tubing
SDP
Supplier:  Thomas Scientific 1158K21
Encompass
Simple, safe, cost effective laboratory waste removal system The Argos MiniVac waste collection system was designed with safety, convenience and bench-top space in mind. The MiniVac system replaces the need for glass waste containers; eliminating the risk of breakage and contamination. The MiniVac flask and cap sit in a safety reservoir basin thatcollects liquid should flask overflow, crack or leak. Full flasks can be transferred safely by using supplied transfer cap. Small footprint (7.5 in.); easy clean up with 70% isopropyl alcohol. The system includes a disposable 1L polystyrene flask, plastic reservoir basin, polypropylene cap with barbed fittings and tubing. Clear PETG plastic safety reservoir collects liquid in case flask cracks or leaks Use as stand-alone waste collection system or overflow vessel Polypropylene barbed cap fittings eliminate risk of broken glass rod fittings Use with in-house vacuum
Catalog No. 50-146-1446