Learn More
Integra Biosciences Corp PLATE HOLDER W/ SLIDE FUNCTION
SDP
Supplier:  Integra Biosciences Corp 6210
Encompass
Plate Holder with slide function For plate reformatting 96 well 384 well
Catalog No. NC1001880