Learn More
Bio Rad Laboratories PLATE LOW-PROFILE 96WELL 25/CS
Supplier:  Bio Rad Laboratories MLL9601
Encompass
PCR Plate, 96-Well Unskirted; Bio-Rad; Multiplate; Low profile for minimized potential condensation; Plates can be cut for use w/fewer samples; PCR volumes of 5-125uL are recommended; Free of DNase, RNase, human DNA; Natural; 25 plates
Catalog No. 50-901-0085