Learn More
Andwin Scientific PLATE ROLLER
SDP
Supplier:  Andwin Scientific RLPLT01
Encompass_Preferred
The Film-Sealing Plate Roller provides secure and even seal for each well. This product is suitable to use with all flat-top multi-well plates.
Catalog No. NC9185793