Learn More
Sigma Aldrich Fine Chemicals Biosciences PlateFugeTM Microplate Mic1EA
Supplier:  Sigma Aldrich Fine Chemicals Biosciences Z7425671EA
Encompass
The Benchmark Scientific C2000 PlateFuge is the first mini sized centrifuge with a swing out rotor for micro plates. Centrifuging plates before PCR ensures all reactants are in the bottom of the wells for proper concentrations and improved yields. In spite of its remarkably small footprint (9x10 inches) the PlateFuge includes a uniquely designed swing out rotor capable of securing 2 micro plates. The easily accessible rotor chamber of the C2000 includes two plate carriers that rest at a 75 degree angle. This allows plates to be inserted confidently without sealing tapes or caps. Upon closing the lid of the centrifuge the rotor accelerates and the centrifugal force swings the plates into a horizontal position. Any droplets on the walls of the plate quickly concentrate into the well bottoms. With a g-force of over 400 xg most samples can be spun down in less than 20 seconds. To end a run simply open the lid and the automatic electric brake brings the rotor to a quick smooth stop i
Catalog No. 50-290-8581