Learn More
Sigma Aldrich Fine Chemicals Biosciences Platelet Derived Growth fa
Supplier:  Sigma Aldrich Fine Chemicals Biosciences P307610UG
Encompass
Platelet-derived growth factor (PDGf) is composed two polypeptide subunits of A and B chain. PDGf is produced by a number of cells including smooth muscle cells endothelial cells and macrophages. The PDGf gene is highly conserved and is mapped to human chromosome 7p22.3. PDGf is structurally and functionally related to vascular endothelial growth factors.
Catalog No. 50-290-4438