Learn More
Apexbio Technology LLC Platelet Membrane Glycoprotein IIB Peptide (296-306) 1mg
SDP
Supplier:  Apexbio Technology LLC A10181
Encompass_Preferred
Platelet Membrane Glycoprotein IIB Peptide 296-306 is a peptide inhibitor targeting glycoprotein IIb (GP IIb) a component of the platelet surface GP IIb-IIIa integrin complex It is designed to disrupt GP IIb-IIIa receptor interactions thereby inhibiting platelet aggregation Platelet Membrane Glycoprotein IIB Peptide 296-306 exerts its biological activity primarily through competitive inhibition of GP IIb binding interfering with platelet adhesion mediated by fibrinogen fibronectin vitronectin or von Willebrand factor Based on these pharmacological properties Platelet Membrane Glycoprotein IIB Peptide 296-306 holds research potential in studies of platelet function thrombosis mechanisms and integrin-mediated signal transduction pathways
Catalog No. 50-000-96455