Learn More
Electron Microscopy Sciences Platinum Blue 1 ML
SDP
Supplier:  Electron Microscopy Sciences 22407
Encompass_Preferred
(IBI Blue Platinum Stain)
CAS #15663-27-1 Pt₄N₈H₆O₂₄C₂0
An EM stain that may have the ability to replace Uranyl Acetate in some instances. If you have issues with having Uranium salts that are radioactive (depleted or not) in the lab this may be the answer for you. An alternative for staining TEM thin sections.
Catalog No. 50-192-6627