Learn More
Electron Microscopy Sciences Platinum Coated Double Edge Razor Blades
SDP
Supplier:  Electron Microscopy Sciences 7200301
Encompass_Preferred
Made from stainless steel, coated with Platinum. Super-sharp-edges. Measures: 62.96 mm (1.69") L x 21.95 mm (0.865") W x 0.10 mm (0.004") Thickness.
Catalog No. 50-949-204