Learn More
Sigma Aldrich Fine Chemicals Biosciences Platinum gauze 100 mesh 991.7G
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 298093
Encompass
Platinum (Pt) is a highly ductile, malleable and unreactive metal. It is resistant to corrosion and stable at high temperatures. Although resistant to hydrochloric and nitric acid, it dissolves readily in hot aqua regia to form chloroplatinic acid.
Catalog No. 50-290-6620