Learn More
Glass Expansion Platinum Skimmer Cone for PerkinElmer Elan 6000/9000/DRCII/DRC-E (WE027803)
SDP
Supplier:  Glass Expansion PE2014PT
Encompass_Preferred
Platinum Skimmer Cone (WE021137) ¿ For PerkinElmer Elan 6000/9000/DRCII/DRC-E ICP-MS. Suitable for most applications, including corrosive samples (e.g., routine aqueous environmental samples, >5% acid matrix, HF, organic). Although the highest cost option, it is typically the most durable, longest-lasting, and because it runs hotter it requires less frequent cleaning. Replace skimmer cone O-ring if brittle, cracked or damaged (NC1585742). Use 10x Magnifier Inspection Tool with Built-In LED Illumination (NC2042261) to inspect cone orifice condition. When not in use, keep cones stored in original packaging for protection.Use skimmer ConeGuard to protect cone threads during cleaning, 70-803-1022. For routine samples, we recommend Nickel Skimmer Cones, see catalog number NC1585740.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-190-7683
 Restricted Item