Learn More
Glass Expansion Platinum Sampler Cone for Agilent 4500/7500 (10mm insert), Suitable for: Agilent 4500, 7500 (G1820-65239)
SDP
Supplier:  Glass Expansion AT1006PT
Encompass_Preferred
Platinum Sampler Cone with 10 mm insert (G1820-65239). For Agilent 4500/7500a/7500i/7500s/7500c/7500ce/7500cs ICP-MS. Suitable for most applications, including corrosive samples (e.g., routine aqueous environmental samples, >5% acid matrix, HF, high TDS). Although the highest cost option, it is typically the most durable, longest-lasting, and because it runs hotter it requires less frequent cleaning. Use 10x Magnifier Inspection Tool with Built-In LED Illumination (NC2042261) to inspect cone orifice condition. Use ConeGuard (70-803-1000) to protect cone threads during cleaning. When not in use, keep cones stored in original packaging for protection.For routine sample, we recommend a nickel sampler cone, see catalog number 501907562.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-190-7569
 Restricted Item