Learn More
Glass Expansion Platinum Sampler cone with Copper Core for Thermo Scientific Element 1/Element 2/Neptune ICP-MS (1067501)
SDP
Supplier:  Glass Expansion TF1006APT/CU
Encompass_Preferred
Platinum Sampler Cone with Copper Core, Boron-Free (1067501) ¿ For Thermo/Finnigan Element1, 2 & Neptune. Excellent for most applications, including corrosive samples (e.g., routine aqueous environmental samples, >5% acid matrix, HF, organic). Although the highest cost option, it is typically the most durable, longest-lasting, and because it runs hotter it requires less frequent cleaning. Use a new Graphite Sampler Cone Gasket (501907717) each time the cone is re-installed, even if it¿s the same cone. Use 10x Magnifier Inspection Tool with Built-In LED Illumination (NC2042261) to inspect cone orifice condition. When not in use, keep cones stored in original packaging for protection.For high sensitivity applications and low matrix containing samples, see catalog number 501907709.For standard Nickel Sampler with Copper Core, see catalog number 501907705.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-190-7710
 Restricted Item