Learn More
Glass Expansion Platinum Skimmer Cone for PerkinElmer NexION 300/350/1000/1100/2000 (W1026907)
SDP
Supplier:  Glass Expansion PE3014PT
Encompass_Preferred
Platinum Skimmer Cone (W1026907) ¿ For PerkinElmer NexION 1000/2000 & 300/350 ICP-MS. Suitable for most applications, including corrosive samples (e.g., routine aqueous environmental samples, >5% acid matrix, HF, organic). Although the highest cost option, it is typically the most durable, longest-lasting, and because it runs hotter it requires less frequent cleaning. Use 10x Magnifier Inspection Tool with Built-In LED Illumination (NC2042261) to inspect cone orifice condition. When not in use, keep cones stored in original packaging for protection. Use skimmer ConeGuard to protect cone threads during cleaning, 70-803-1026.For routine samples, we recommend Nickel Skimmer Cones, see catalog number 501907685. For lowest boron background, we recommend Boron-Free Platinum Sampler Cones, see catalog number 501907689.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-190-7688
 Restricted Item