Learn More
Glass Expansion Platinum Skimmer Cone with Nickel Base for x-lens for Agilent 7900/8900 ICP-MS (G3280-67202)
SDP
Supplier:  Glass Expansion AT7908XPT/NI
Encompass_Preferred
Platinum Skimmer Cone with Nickel Base for x-lens (G3280-67202) ¿ For Agilent 7900/8900 ICP-MS. Recommended for organic samples. More durable, longer-lasting, & because it runs hotter it requires less frequent cleaning. Use 10x Magnifier Inspection Tool with Built-In LED Illumination (NC2042261) to inspect cone orifice condition. Use Skimmer ConeGuard (70-803-1004) to protect cone threads during cleaning. When not in use, keep cones stored in original packaging for protection.For routine applications, use standard Nickel Skimmer for x-lens, see P/N 501907593.For corrosive analysis, use Platinum Skimmer with Cu Base for x-lens, P/N 501907596, which must be mounted on brass adaptor skimmer base, see P/N AT7905X-BR. For lowest nickel background and laser ablation applications, use Aluminum Skimmer for x-lens, see P/N AT7902X-Al.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-190-7595