Learn More
XRF Scientific Americas Inc. PLATINUM TIPPED 5 GRAMS STAINLESS STEEL TONG 300MM LONG
Supplier:  XRF Scientific Americas Inc. TGS3005
Encompass
Saintless steel tong with platinum tips 300 mm long. These tongs are used to manipulate platinum labware and avoid contamination on the platinum crucibles or dishes.
Catalog No. NC1231733