Learn More
Sigma Aldrich Fine Chemicals Biosciences Platinum wire diam. 0.5 mm2G
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 267201
Encompass
Platinum (Pt) is a highly ductile, malleable and unreactive metal. It is resistant to corrosion and stable at high temperatures. Although resistant to hydrochloric and nitric acid, it dissolves readily in hot aqua regia to form chloroplatinic acid.
Catalog No. 50-290-6715