Learn More
Sigma Aldrich Fine Chemicals Biosciences Platinum wire diam. 2.0 mm
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 444685
Encompass
Platinum (Pt) wire is a metal wire that is a biocompatible material that can be used in electrodes and implants. The efficiency of the platinum electrode can be improved by forming a platinum black surface.
Catalog No. 50-290-6792
 Restricted Item