Learn More
Medchemexpress LLC Plerixafor | 110078-46-1 | MFCD05662218 | 99.9% | 502.78 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY10046
Encompass_Preferred
Plerixafor (AMD 3100) is a selective CXCR4 antagonist and an allosteric agonist of CXCR7. It functions as an immunostimulant and a hematopoietic stem cell (HSC) mobilizer. This compound inhibits HIV-1 and HIV-2 replication at low nanomolar concentrations.
  • Functions as an immunostimulant.
  • Mobilizes hematopoietic stem cells.
  • Inhibits HIV-1 and HIV-2 replication.
  • Acts as a selective CXCR4 antagonist.
  • Allosteric agonist of CXCR7.
Catalog No. 50-003-47515