Learn More
Medchemexpress LLC Plerixafor | 110078-46-1 | MFCD05662218 | 99.9% | 502.78 g/mol | C28H54N8 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY1004650MG
Encompass_Preferred
Plerixafor is a small-molecule CXCR4 antagonist used in research to mobilize hematopoietic stem cells and to probe CXCR4-mediated signaling pathways. It is provided as a solid reagent for in vitro and ex vivo studies and has reported antiviral activity in cell-based assays.
  • Selective CXCR4 antagonist with reported IC50 ≈ 44 nM
  • Mobilizes hematopoietic stem cells for ex vivo applications
  • Useful tool for CXCR4 pathway, chemotaxis, and signaling studies
  • Documented in vitro antiviral activity against HIV strains
  • Supplied as a powder suitable for laboratory reagent use
  • High reported purity and well-defined chemical identity
Catalog No. 50-002-14085