Learn More
TARGETMOL CHEMICALS INC Plerixafor 25MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T177625MG
Encompass_Preferred
Also available in 5 mg, 10 mg, 50 mg, 100 mg, 200 mg, 500 mg and bulk. Please contact Fisher for quotes. Plerixafor (AMD-3329), a chemokine receptor antagonist, blocks the binding of stromal cell-derived factor (SDF-1alpha) to the cellular receptor CXCR4. Purity 100%
Catalog No. 50-287-8314