Learn More
Selleck Chemical LLC Plerixafor (AMD3100) 8HCl S3013-1g
SDP
Supplier:  Selleck Chemical LLC S30131G
Encompass_Preferred
Plerixafor (AMD3100 JM 3100 Plerixafor Octahydrochloride AMD3100 octahydrochloride SID791 octahydrochloride) 8HCl is the hydrochloride of Plerixafor a chemokine receptor antagonist for CXCR4 and CXCL12-mediated chemotaxis with IC50 of 44 nM and 5 7 nM in cell-free assays respectively Plerixafor can be used as an anti-HIV agent
Catalog No. 50-311-0737